Anti-XRCC1 Mouse Monoclonal Antibody [clone: OTI3C3]

Lieferant: OriGene
TA500847
ORIGTA500847EA 621 EUR
ORIGTA500847
Anti-XRCC1 Mouse Monoclonal Antibody [clone: OTI3C3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: XRCC1 (X-ray repair complementing defective repair in Chinese hamster cells 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR