Anti-LOX Mouse Monoclonal Antibody [clone: OTI9A11]

Lieferant: OriGene
TA500877
ORIGTA500877EA 621 EUR
ORIGTA500877
Anti-LOX Mouse Monoclonal Antibody [clone: OTI9A11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR