Anti-SHC1 Mouse Monoclonal Antibody [clone: OTI1H8]

Lieferant: OriGene
TA501075
ORIGTA501075EA 621 EUR
ORIGTA501075
Anti-SHC1 Mouse Monoclonal Antibody [clone: OTI1H8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR