Anti-NIT2 Mouse Monoclonal Antibody [clone: OTI3F3]

Lieferant: OriGene
TA501098
ORIGTA501098EA 621 EUR
ORIGTA501098
Anti-NIT2 Mouse Monoclonal Antibody [clone: OTI3F3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NIT2 (nitrilase family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR