Anti-BTK Mouse Monoclonal Antibody [clone: OTI1H7]

Lieferant: OriGene
TA501101
ORIGTA501101EA 621 EUR
ORIGTA501101
Anti-BTK Mouse Monoclonal Antibody [clone: OTI1H7]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: BTK (Bruton agammaglobulinemia tyrosine kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR