Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI2E9]

Lieferant: OriGene
TA501197
ORIGTA501197EA 621 EUR
ORIGTA501197
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI2E9]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2 (cytochrome P450, family 1, subfamily A, polypeptide 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR