Anti-BSG Mouse Monoclonal Antibody [clone: OTI4E4]

Lieferant: OriGene
TA501212
ORIGTA501212EA 621 EUR
ORIGTA501212
Anti-BSG Mouse Monoclonal Antibody [clone: OTI4E4]
Antikörper
  • Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR