Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI3A11]

Lieferant: OriGene
TA501217
ORIGTA501217EA 621 EUR
ORIGTA501217
Anti-ACAT2 Mouse Monoclonal Antibody [clone: OTI3A11]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2 (acetyl-CoA acetyltransferase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR