Anti-C9orf41 Mouse Monoclonal Antibody [clone: OTI4A2]

Lieferant: OriGene
TA501298
ORIGTA501298EA 621 EUR
ORIGTA501298
Anti-C9orf41 Mouse Monoclonal Antibody [clone: OTI4A2]
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: C9orf41 (chromosome 9 open reading frame 41)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR