Anti-EIF2S1 Mouse Monoclonal Antibody [clone: OTI3H4]

Lieferant: OriGene
TA501313
ORIGTA501313EA 621 EUR
ORIGTA501313
Anti-EIF2S1 Mouse Monoclonal Antibody [clone: OTI3H4]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EIF2S1 (eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR