Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1C1]

Lieferant: OriGene
TA501479
ORIGTA501479EA 621 EUR
ORIGTA501479
Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1C1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LIN7B (lin-7 homolog B (C. elegans))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR