Anti-DYNC1LI1 Mouse Monoclonal Antibody [clone: OTI2E8]

Lieferant: OriGene
TA501509
ORIGTA501509EA 621 EUR
ORIGTA501509
Anti-DYNC1LI1 Mouse Monoclonal Antibody [clone: OTI2E8]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DYNC1LI1 (dynein, cytoplasmic 1, light intermediate chain 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR