Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1D12]

Lieferant: OriGene
TA501554
ORIGTA501554EA 621 EUR
ORIGTA501554
Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1D12]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LIN7B (lin-7 homolog B (C. elegans))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR