Anti-BEST3 Mouse Monoclonal Antibody [clone: OTI2H3]

Lieferant: OriGene
TA501979
ORIGTA501979EA 621 EUR
ORIGTA501979
Anti-BEST3 Mouse Monoclonal Antibody [clone: OTI2H3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BEST3 (bestrophin 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR