Anti-SPR Mouse Monoclonal Antibody [clone: OTI1F4]

Lieferant: OriGene
TA501984
ORIGTA501984EA 621 EUR
ORIGTA501984
Anti-SPR Mouse Monoclonal Antibody [clone: OTI1F4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR