Anti-PBLD Mouse Monoclonal Antibody [clone: OTI7G5]

Lieferant: OriGene
TA501994
ORIGTA501994EA 621 EUR
ORIGTA501994
Anti-PBLD Mouse Monoclonal Antibody [clone: OTI7G5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PBLD (phenazine biosynthesis-like protein domain containing)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR