Anti-NHEJ1 Mouse Monoclonal Antibody [clone: OTI1G7]

Lieferant: OriGene
TA502008
ORIGTA502008EA 621 EUR
ORIGTA502008
Anti-NHEJ1 Mouse Monoclonal Antibody [clone: OTI1G7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NHEJ1 (nonhomologous end-joining factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR