Anti-VSIG2 Mouse Monoclonal Antibody [clone: OTI1H2]

Lieferant: OriGene
TA502124
ORIGTA502124EA 621 EUR
ORIGTA502124
Anti-VSIG2 Mouse Monoclonal Antibody [clone: OTI1H2]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: VSIG2 (V-set and immunoglobulin domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR