Anti-DNAJA2 Mouse Monoclonal Antibody [clone: OTI1G9]

Lieferant: OriGene
TA502126
ORIGTA502126EA 621 EUR
ORIGTA502126
Anti-DNAJA2 Mouse Monoclonal Antibody [clone: OTI1G9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJA2 (DnaJ (Hsp40) homolog, subfamily A, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR