Anti-BIN3 Mouse Monoclonal Antibody [clone: OTI8E9]

Lieferant: OriGene
TA502145
ORIGTA502145EA 621 EUR
ORIGTA502145
Anti-BIN3 Mouse Monoclonal Antibody [clone: OTI8E9]
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BIN3 (bridging integrator 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR