Anti-HOXC11 Mouse Monoclonal Antibody [clone: OTI4H8]

Lieferant: OriGene
TA502578
ORIGTA502578EA 621 EUR
ORIGTA502578
Anti-HOXC11 Mouse Monoclonal Antibody [clone: OTI4H8]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: HOXC11 (homeobox C11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR