Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI1A8]

Lieferant: OriGene
TA502585
ORIGTA502585EA 621 EUR
ORIGTA502585
Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI1A8]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LACTB2 (lactamase, beta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR