Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI5F7]

Lieferant: OriGene
TA502829
ORIGTA502829EA 621 EUR
ORIGTA502829
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI5F7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR