Anti-PPM1B Mouse Monoclonal Antibody [clone: OTI3A10]

Lieferant: OriGene
TA502862
ORIGTA502862EA 621 EUR
ORIGTA502862
Anti-PPM1B Mouse Monoclonal Antibody [clone: OTI3A10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PPM1B (protein phosphatase, Mg2+/Mn2+ dependent, 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR