Anti-PTPN1 Mouse Monoclonal Antibody [clone: OTI1B4]

Lieferant: OriGene
TA503037
ORIGTA503037EA 621 EUR
ORIGTA503037
Anti-PTPN1 Mouse Monoclonal Antibody [clone: OTI1B4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PTPN1 (protein tyrosine phosphatase, non-receptor type 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR