Anti-LCMT1 Mouse Monoclonal Antibody [clone: OTI1E8]

Lieferant: OriGene
TA503126
ORIGTA503126EA 621 EUR
ORIGTA503126
Anti-LCMT1 Mouse Monoclonal Antibody [clone: OTI1E8]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: LCMT1 (leucine carboxyl methyltransferase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR