Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI4D6]

Lieferant: OriGene
TA503136
ORIGTA503136EA 621 EUR
ORIGTA503136
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI4D6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR