Anti-UBOX5 Mouse Monoclonal Antibody [clone: OTI5A2]

Lieferant: OriGene
TA503202
ORIGTA503202EA 621 EUR
ORIGTA503202
Anti-UBOX5 Mouse Monoclonal Antibody [clone: OTI5A2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: UBOX5 (U-box domain containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR