Anti-TOMM34 Mouse Monoclonal Antibody [clone: OTI1G7]

Lieferant: OriGene
TA503216
ORIGTA503216EA 621 EUR
ORIGTA503216
Anti-TOMM34 Mouse Monoclonal Antibody [clone: OTI1G7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TOMM34 (translocase of outer mitochondrial membrane 34)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR