Anti-ERCC4 Mouse Monoclonal Antibody [clone: OTI4E11]

Lieferant: OriGene
TA503261
ORIGTA503261EA 621 EUR
ORIGTA503261
Anti-ERCC4 Mouse Monoclonal Antibody [clone: OTI4E11]
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC4 (excision repair cross-complementation group 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR