Anti-SETD7 Mouse Monoclonal Antibody [clone: OTI2D10]

Lieferant: OriGene
TA503322
ORIGTA503322EA 621 EUR
ORIGTA503322
Anti-SETD7 Mouse Monoclonal Antibody [clone: OTI2D10]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SETD7 (SET domain containing (lysine methyltransferase) 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR