Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI2B7]

Lieferant: OriGene
TA503616
ORIGTA503616EA 621 EUR
ORIGTA503616
Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI2B7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NIF3L1 (NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR