Anti-APMAP Mouse Monoclonal Antibody [clone: OTI4F6]

Lieferant: OriGene
TA504220
ORIGTA504220EA 621 EUR
ORIGTA504220
Anti-APMAP Mouse Monoclonal Antibody [clone: OTI4F6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: APMAP (adipocyte plasma membrane associated protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR