Anti-EPN2 Mouse Monoclonal Antibody [clone: OTI1F5]

Lieferant: OriGene
TA504301
ORIGTA504301EA 621 EUR
ORIGTA504301
Anti-EPN2 Mouse Monoclonal Antibody [clone: OTI1F5]
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: EPN2 (epsin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR