Anti-REEP2 Mouse Monoclonal Antibody [clone: OTI2B7]

Lieferant: OriGene
TA504322
ORIGTA504322EA 621 EUR
ORIGTA504322
Anti-REEP2 Mouse Monoclonal Antibody [clone: OTI2B7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: REEP2 (receptor accessory protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR