Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI12F11]

Lieferant: OriGene
TA504333
ORIGTA504333EA 621 EUR
ORIGTA504333
Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI12F11]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR