Anti-FMR1 Mouse Monoclonal Antibody [clone: OTI1H9]

Lieferant: OriGene
TA504342
ORIGTA504342EA 621 EUR
ORIGTA504342
Anti-FMR1 Mouse Monoclonal Antibody [clone: OTI1H9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR