Anti-S100P Mouse Monoclonal Antibody [clone: OTI1A4]

Lieferant: OriGene
TA504478
ORIGTA504478EA 621 EUR
ORIGTA504478
Anti-S100P Mouse Monoclonal Antibody [clone: OTI1A4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: S100P (S100 calcium binding protein P)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR