Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C11]

Lieferant: OriGene
TA504493
ORIGTA504493EA 621 EUR
ORIGTA504493
Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR