Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI7G2]

Lieferant: OriGene
TA504695
ORIGTA504695EA 621 EUR
ORIGTA504695
Anti-CPA2 Mouse Monoclonal Antibody [clone: OTI7G2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CPA2 (carboxypeptidase A2 (pancreatic))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR