Anti-TBCC Mouse Monoclonal Antibody [clone: OTI2B3]

Lieferant: OriGene
TA504709
ORIGTA504709EA 621 EUR
ORIGTA504709
Anti-TBCC Mouse Monoclonal Antibody [clone: OTI2B3]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-4000; Immunochemistry: 1:150

Type: Primary
Antigen: TBCC (tubulin folding cofactor C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR