Anti-MPI Mouse Monoclonal Antibody [clone: OTI4G7]

Lieferant: OriGene
TA504772
ORIGTA504772EA 621 EUR
ORIGTA504772
Anti-MPI Mouse Monoclonal Antibody [clone: OTI4G7]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MPI (mannose phosphate isomerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol MPI
Environmentally Preferable
Antigen-Name mannose phosphate isomerase
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Klon OTI4G7
Reaktivität Human
Wirt Mouse
Isotyp IgG1
Western blot Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Durchflusszytometrie Yes
Epitop Full length
Form Liquid
Molecular weight 46.5 kDa
Reinigung purified by affinity chromatography
Kreuzadsorption none
Lagerungspuffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR