Anti-APCS Mouse Monoclonal Antibody [clone: OTI1D6]

Lieferant: OriGene
TA505247
ORIGTA505247EA 621 EUR
ORIGTA505247
Anti-APCS Mouse Monoclonal Antibody [clone: OTI1D6]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR