Anti-UBE2G2 Mouse Monoclonal Antibody [clone: OTI1D7]

Lieferant: OriGene
TA505258
ORIGTA505258EA 621 EUR
ORIGTA505258
Anti-UBE2G2 Mouse Monoclonal Antibody [clone: OTI1D7]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: UBE2G2 (ubiquitin-conjugating enzyme E2G 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR