Anti-SRY Mouse Monoclonal Antibody [clone: OTI3C8]

Lieferant: OriGene
TA505267
ORIGTA505267EA 621 EUR
ORIGTA505267
Anti-SRY Mouse Monoclonal Antibody [clone: OTI3C8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SRY (sex determining region Y)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR