Anti-CMPK1 Mouse Monoclonal Antibody [clone: OTI1A1]

Lieferant: OriGene
TA505389
ORIGTA505389EA 621 EUR
ORIGTA505389
Anti-CMPK1 Mouse Monoclonal Antibody [clone: OTI1A1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CMPK1 (cytidine monophosphate (UMP-CMP) kinase 1, cytosolic)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR