Anti-KIF25 Mouse Monoclonal Antibody [clone: OTI2G9]

Lieferant: OriGene
TA505429
ORIGTA505429EA 621 EUR
ORIGTA505429
Anti-KIF25 Mouse Monoclonal Antibody [clone: OTI2G9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KIF25 (kinesin family member 25)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR