Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: OTI1B10]

Lieferant: OriGene
TA505501
ORIGTA505501EA 621 EUR
ORIGTA505501
Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: OTI1B10]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: TRAPPC4 (trafficking protein particle complex 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR