Anti-TRMT2A Mouse Monoclonal Antibody [clone: OTI3D8]

Lieferant: OriGene
TA505517
ORIGTA505517EA 621 EUR
ORIGTA505517
Anti-TRMT2A Mouse Monoclonal Antibody [clone: OTI3D8]
Antikörper
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TRMT2A (tRNA methyltransferase 2 homolog A (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR