Anti-GADD45G Mouse Monoclonal Antibody [clone: OTI5G2]

Lieferant: OriGene
TA505535
ORIGTA505535EA 621 EUR
ORIGTA505535
Anti-GADD45G Mouse Monoclonal Antibody [clone: OTI5G2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GADD45G (growth arrest and DNA-damage-inducible, gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR